9-Phenylcarbazole-2-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (9-Phenyl-9H-carbazol-2-yl)boronic acid

A180803-100G A180803-100MG A180803-10G A180803-1G A180803-250MG A180803-25G
76824-886EA 203 USD
76824-886 76824-888 76824-890 76824-892 76824-894 76824-896
9-Phenylcarbazole-2-boronic acid 98%
9-Phenylcarbazole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₄BNO₂
MDL Number: MFCD22207050
CAS Number: 1001911-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR