2-Hydroxy-3-methyl-2-cyclopentenone 95%

Supplier: Ambeed
Warning

Synonyms: Cyclotene, 2-Hydroxy-3-methylcyclopent-2-enone, 3-Methyl-2-cyclopentenon-2-ol, Methyl cyclopentenolone

A180552-100G A180552-25G A180552-5G
76824-872EA 22.28 USD
76824-872 76824-874 76824-876
2-Hydroxy-3-methyl-2-cyclopentenone 95%
2-Hydroxy-3-methyl-2-cyclopentenone

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00013747
CAS Number: 80-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR