Ethyl 2-bromo-6-fluorobenzoate 98%

Supplier: Ambeed
Warning

A180258-100MG A180258-1G A180258-250MG
76824-844EA 8.49 USD
76824-844 76824-846 76824-848
Ethyl 2-bromo-6-fluorobenzoate 98%
Ethyl 2-bromo-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
MDL Number: MFCD10000924
CAS Number: 1214362-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR