3-Bromo-5-(4-morpholino)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Bromo-3-pyridyl)morpholine, 3-Bromo-5-morpholinopyridine

A180215-100MG A180215-1G A180215-250MG A180215-5G
76824-834EA 15.92 USD
76824-834 76824-836 76824-838 76824-840
3-Bromo-5-(4-morpholino)pyridine 98%
3-Bromo-5-(4-morpholino)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
MDL Number: MFCD11656285
CAS Number: 200064-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR