Benzyl (3-aminopropyl)carbamate 95%

Supplier: Ambeed
Warning

A180023-10G A180023-1G A180023-25G A180023-250MG A180023-5G A180023-100G
76824-830EA 388.29 USD
76824-830 77593-146 77611-744 77683-444 77683-445 77683-443
Benzyl (3-aminopropyl)carbamate 95%
Benzyl (3-aminopropyl)carbamate

Precaución: For research use only.

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD02094511
CAS Number: 46460-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR