3-Amino-6-bromopyridin-2(1H)-one hydrobromide 97%

Supplier: Ambeed
Warning

A179900-100MG A179900-1G A179900-250MG
76824-824EA 17 USD
76824-824 76824-826 76824-828
3-Amino-6-bromopyridin-2(1H)-one hydrobromide 97%
3-Amino-6-bromopyridin-2(1H)-one hydrobromide

Precaución: for Research Use Only

Formula: C₅H₆Br₂N₂O
MDL Number: MFCD09701347
CAS Number: 1187930-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR