Ethyl 3-(2,5-dichlorophenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A179834-1G A179834-250MG
76824-820EA 20 USD
76824-820 76824-822
Ethyl 3-(2,5-dichlorophenyl)-3-oxopropanoate 95%
Ethyl 3-(2,5-dichlorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂O₃
MDL Number: MFCD08702767
CAS Number: 53090-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR