2-Bromo-N-methylthiazole-4-carboxamide 98%

Supplier: Ambeed
Danger

A178805-1G A178805-250MG
76824-812EA 148.53 USD
76824-812 76824-814
2-Bromo-N-methylthiazole-4-carboxamide 98%
2-Bromo-N-methylthiazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₅H₅BrN₂OS
MW: 221.07 g/mol
MDL Number: MFCD20265433
CAS Number: 1092942-42-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR