1-Chloroethylcyclohexyl carbonate 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A177801-25G A177801-100G A177801-500G
76824-800EA 11 USD
76824-800 77683-421 77885-378
1-Chloroethylcyclohexyl carbonate 97%, Ambeed.Inc
1-Chloroethylcyclohexyl carbonate

Precaución: For research use only.

Formula: C₉H₁₅ClO₃
MDL Number: MFCD04038149
CAS Number: 99464-83-2
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR