Piperazin-2-one, N1-BOC protected 95%

Supplier: Ambeed
Warning

A177754-100MG A177754-1G A177754-250MG
76824-792EA 44 USD
76824-792 76824-794 76824-796
Piperazin-2-one, N1-BOC protected 95%
Piperazin-2-one, N1-BOC protected

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
MDL Number: MFCD06797597
CAS Number: 889958-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR