5'-Chloro-2'-hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A177618-10G A177618-1G A177618-250MG A177618-5G
76824-784EA 222.79 USD
76824-784 76824-786 76824-788 76824-790
5'-Chloro-2'-hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid 95%
5'-Chloro-2'-hydroxy-3'-nitro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈ClNO₅
MW: 293.66 g/mol
MDL Number: MFCD13184302
CAS Number: 376592-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR