1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 2-hydroxypropanoic acid salt 97%

Supplier: Ambeed
Warning

A177117-100G A177117-1G A177117-250MG A177117-25G A177117-500G A177117-5G
76824-758EA 466.8 USD
76824-758 76824-760 76824-762 76824-764 76824-766 76824-768
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 2-hydroxypropanoic acid salt 97%
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 2-hydroxypropanoic acid salt

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₃O₆
MW: 421.41 g/mol
MDL Number: MFCD01759747
CAS Number: 97867-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR