N,2-Dihydroxy-5-nitrobenzamide 95%
Supplier: Ambeed
|
Warning
|
A177037-250MG
A177037-5G
A177037-1G
76824-750EA
18.04
USD
76824-750
76824-752
77683-417
N,2-Dihydroxy-5-nitrobenzamide 95%
N,2-Dihydroxy-5-nitrobenzamide
Precaución: For research use only.
Formula:
C₇H₆N₂O₅ MW: 198.13 g/mol |
MDL Number:
MFCD00818219 CAS Number: 61494-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...