1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A176983-100MG A176983-250MG A176983-5G
76824-744EA 50.93 USD
76824-744 76824-746 76824-748
1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid 97%
1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrFO₂
MW: 259.072 g/mol
MDL Number: MFCD12405622
CAS Number: 872422-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR