(Z)-2-Methyl-4-(4-methylbenzylidene)oxazol-5(4H)-one 95%

Supplier: Ambeed
Warning

A176916-100MG A176916-1G A176916-250MG
76824-738EA 23.34 USD
76824-738 76824-740 76824-742
(Z)-2-Methyl-4-(4-methylbenzylidene)oxazol-5(4H)-one 95%
(Z)-2-Methyl-4-(4-methylbenzylidene)oxazol-5(4H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MW: 201.22 g/mol
MDL Number: MFCD00220066
CAS Number: 93634-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR