1-Benzyl-2-(chloromethyl)benzene 97%

Supplier: Ambeed
Danger

A176792-100MG A176792-250MG
76824-734EA 13.8 USD
76824-734 76824-736
1-Benzyl-2-(chloromethyl)benzene 97%
1-Benzyl-2-(chloromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl
MW: 216.71 g/mol
MDL Number: MFCD12025181
CAS Number: 7510-28-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR