3,4,5-Trimethoxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A175709-1G A175709-5G
76824-710EA 76 USD
76824-710 76824-712
3,4,5-Trimethoxyphenylboronic acid pinacol ester 98%
3,4,5-Trimethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₅
MDL Number: MFCD12405526
CAS Number: 214360-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR