2-(1H-Imidazol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98%

Supplier: Ambeed
Warning

A175654-100MG A175654-1G A175654-250MG A175654-5G
76824-702EA 37.14 USD
76824-702 76824-704 76824-706 76824-708
2-(1H-Imidazol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98%
2-(1H-Imidazol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₄O₂
MW: 272.11 g/mol
MDL Number: MFCD11878350
CAS Number: 1160790-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR