Glycine hydrochloride (H-Gly-OH.HCl) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Gly-OH·HCl, Glycine hydrochloride
A175581-100G
A175581-500G
A175581-1000G
A175581-25G
76824-698EA
19
USD
76824-698
76824-700
77885-367
77885-368
Glycine hydrochloride (H-Gly-OH.HCl) 99%
Glycine hydrochloride (H-Gly-OH.HCl)
Precaución: for Research Use Only
Formula:
C₂H₆ClNO₂ MW: 111.53 g/mol |
MDL Number:
MFCD00012872 CAS Number: 6000-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...