5-Bromo-2-iodo-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-iodo-4-picoline
A175276-100MG
A175276-250MG
A175276-1G
76824-674EA
38.2
USD
76824-674
76824-676
77683-407
5-Bromo-2-iodo-4-methylpyridine 98%
5-Bromo-2-iodo-4-methylpyridine
Precaución: For research use only.
Formula:
C₆H₅BrIN MW: 297.92 g/mol |
MDL Number:
MFCD09801049 CAS Number: 941294-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...