tert-Butyl-3-(isopropyl)carbazate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-tert-Butoxycarbonyl-2-isopropylhydrazine, 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-Boc-2-isopropylhydrazine, tert-Butyl 2-isopropylhydrazinecarboxylate
A175197-100G
A175197-10G
A175197-1G
A175197-250MG
A175197-25G
A175197-5G
76824-662EA
435
USD
76824-662
76824-664
76824-666
76824-668
76824-670
76824-672
tert-Butyl-3-(isopropyl)carbazate 95%
tert-Butyl-3-(isopropyl)carbazate
Precaución: for Research Use Only
Formula:
C₈H₁₈N₂O₂ MW: 174.24 g/mol |
MDL Number:
MFCD14635748 CAS Number: 16689-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...