Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate hydrochloride 96%

Supplier: Ambeed
Warning

A175006-100G A175006-10G A175006-1G A175006-250MG A175006-25G A175006-5G
76824-646EA 662 USD
76824-646 76824-648 76824-650 76824-652 76824-654 76824-656
Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate hydrochloride 96%
Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄ClNO₆
MDL Number: MFCD18206207
CAS Number: 183322-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR