3-Bromoquinolin-6-ol 97%

Supplier: Ambeed
Warning

A174499-100MG A174499-1G A174499-250MG
76824-620EA 22 USD
76824-620 76824-622 76824-624
3-Bromoquinolin-6-ol 97%
3-Bromoquinolin-6-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD18260430
CAS Number: 13669-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR