Methyl 4-Bromo-2-chlorobenzoate 98%

Supplier: Ambeed
Warning

A174308-10G A174308-1G A174308-250MG A174308-25G A174308-5G
76824-608EA 29 USD
76824-608 76824-610 76824-612 76824-614 76824-616
Methyl 4-Bromo-2-chlorobenzoate 98%
Methyl 4-Bromo-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD10566823
CAS Number: 185312-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR