4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A173951-1G A173951-250MG A173951-5G
76824-602EA 90.18 USD
76824-602 76824-604 76824-606
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine 98%
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₂
MW: 191.04 g/mol
MDL Number: MFCD04039039
CAS Number: 869901-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR