1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chlorophenyl)-1H-pyrazol-3-ol

A173662-100G A173662-10G A173662-5G A173662-25G A173662-500G
76824-586EA 18 USD
76824-586 76824-588 76824-590 77082-422 77885-353
1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole 98%
1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD08459260
CAS Number: 76205-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR