Methyl 5-amino-2,6-dichloropyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A171880-100MG A171880-10G A171880-250MG A171880-1G A171880-5G
76824-532EA 31 USD
76824-532 76824-534 76824-536 77885-346 77885-347
Methyl 5-amino-2,6-dichloropyrimidine-4-carboxylate 95%
Methyl 5-amino-2,6-dichloropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N₃O₂
MDL Number: MFCD16660538
CAS Number: 502184-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR