6-Fluoro-2,3-dimethoxybenzoic acid 95%

Supplier: Ambeed
Warning

A171474-100MG A171474-250MG
76824-510EA 27 USD
76824-510 76824-512
6-Fluoro-2,3-dimethoxybenzoic acid 95%
6-Fluoro-2,3-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₄
MDL Number: MFCD13193706
CAS Number: 265670-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR