tert-Butyl 4-bromo-1H-indazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A170906-100MG A170906-1G A170906-250MG A170906-5G
76824-490EA 14 USD
76824-490 76824-492 76824-494 76824-496
tert-Butyl 4-bromo-1H-indazole-5-carboxylate 97%
tert-Butyl 4-bromo-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MDL Number: MFCD22572308
CAS Number: 1203662-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR