1-Bromo-2,6-difluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1,3-difluoro-5-nitrobenzene

A170560-1G A170560-250MG A170560-25G A170560-5G
76824-474EA 33.95 USD
76824-474 76824-476 76824-478 76824-480
1-Bromo-2,6-difluoro-4-nitrobenzene 98%
1-Bromo-2,6-difluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.9864 g/mol
MDL Number: MFCD04974121
CAS Number: 886762-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR