(2S,5R,6S)-Benzhydryl 6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%

Supplier: Ambeed
Warning

A169898-250MG A169898-5G
76824-460EA 24 USD
76824-460 76824-462
(2S,5R,6S)-Benzhydryl 6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%
(2S,5R,6S)-Benzhydryl 6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide

Precaución: for Research Use Only

Formula: C₂₁H₂₀BrNO₄S
MDL Number: MFCD21608586
CAS Number: 80353-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR