4-Fluoronaphthalen-1-ol 95%
Supplier: Ambeed
|
Warning
|
A169771-100MG
A169771-1G
A169771-5G
A169771-250MG
76824-450EA
19.1
USD
76824-450
76824-452
76824-454
77683-388
4-Fluoronaphthalen-1-ol 95%
4-Fluoronaphthalen-1-ol
Precaución: For research use only.
Formula:
C₁₀H₇FO |
MDL Number:
MFCD10699695 CAS Number: 315-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...