Methyl 3-bromo-5-formylbenzoate 98%

Supplier: Ambeed
Warning

A169514-100MG A169514-1G A169514-250MG A169514-5G
76824-448EA 24 USD
76824-448 77885-335 77885-336 77885-337
Methyl 3-bromo-5-formylbenzoate 98%
Methyl 3-bromo-5-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MDL Number: MFCD22381229
CAS Number: 377734-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR