(5-Bromobenzofuran-2-yl)boronic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A169114-250MG A169114-1G A169114-5G A169114-100MG A169114-25G
76824-444EA 16 USD
76824-444 77593-042 77593-044 77683-385 77885-331
(5-Bromobenzofuran-2-yl)boronic acid 97%, Ambeed.Inc
(5-Bromobenzofuran-2-yl)boronic acid

Precaución: For research use only.

Formula: C₈H₆BBrO₃
MW: 240.846 g/mol
MDL Number: MFCD06801726
CAS Number: 331833-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR