3-(Methylsulfonyl)benzyl alcohol 96%

Supplier: Ambeed
Warning

Synonyms: (3-(Methylsulphonyl)phenyl)methanol

A169001-1G A169001-250MG
76824-434EA 198.39 USD
76824-434 76824-436
3-(Methylsulfonyl)benzyl alcohol 96%
3-(Methylsulfonyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃S
MW: 186.228 g/mol
MDL Number: MFCD15527275
CAS Number: 220798-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR