6-Bromo-4-chloropyridin-2-amine 95%

Supplier: Ambeed
Warning

A168893-100MG A168893-250MG A168893-1G A168893-5G
76824-424EA 82.76 USD
76824-424 76824-426 77550-086 77567-478
6-Bromo-4-chloropyridin-2-amine 95%
6-Bromo-4-chloropyridin-2-amine

Precaución: For research use only.

Formula: C₅H₄BrClN₂
MW: 207.45 g/mol
MDL Number: MFCD14582019
CAS Number: 1206249-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR