6-Bromo-o-anisaldehyde 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methoxybenzaldehyde

A168889-100MG A168889-5G A168889-1G A168889-250MG
76824-420EA 17 USD
76824-420 76824-422 77611-644 77683-381
6-Bromo-o-anisaldehyde 95%, Ambeed.Inc
6-Bromo-o-anisaldehyde

Precaución: For research use only.

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD08059232
CAS Number: 126712-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR