(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-N-t-Boc-3-amino-1,2-epoxy-4-phenylbutane, (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, Darunavir Impurity 22

A168873-1G A168873-250MG A168873-500G A168873-10G A168873-25G
76824-414EA 11.67 USD
76824-414 76824-416 76824-418 77550-084 77567-474
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane 98%
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane

Precaución: For research use only.

Formula: C₁₅H₂₁NO₃
MW: 263.34 g/mol
MDL Number: MFCD02258997
CAS Number: 98737-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR