6-Chloro-5-(trifluoromethyl)picolinic acid 95%

Supplier: Ambeed
Warning

A168344-100MG A168344-1G A168344-250MG A168344-5G
76824-396EA 64.72 USD
76824-396 76824-398 76824-400 76824-402
6-Chloro-5-(trifluoromethyl)picolinic acid 95%
6-Chloro-5-(trifluoromethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
MDL Number: MFCD18257438
CAS Number: 855915-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR