1-(2,6-Difluoropyridin-3-yl)ethanone 95%

Supplier: Ambeed
Danger

A167808-1G A167808-250MG A167808-5G A167808-100MG
76824-384EA 39 USD
76824-384 76824-386 76824-388 77885-327
1-(2,6-Difluoropyridin-3-yl)ethanone 95%
1-(2,6-Difluoropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅F₂NO
MDL Number: MFCD11520715
CAS Number: 920036-27-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR