2,4-Dichlorofuro[3,4-d]pyrimidin-7(5H)-one 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A167657-1G A167657-250MG A167657-5G A167657-100MG
76824-376EA 88 USD
76824-376 76824-378 76824-380 77683-374
2,4-Dichlorofuro[3,4-d]pyrimidin-7(5H)-one 95%, Ambeed.Inc
2,4-Dichlorofuro[3,4-d]pyrimidin-7(5H)-one

Precaución: For research use only.

Formula: C₆H₂Cl₂N₂O₂
MW: 205.00 g/mol
MDL Number: MFCD18072502
CAS Number: 15783-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR