Methyl 6-bromo-3-methoxypicolinate 98%

Supplier: Ambeed
Warning

A167550-100MG A167550-250MG A167550-5G
76824-368EA 20 USD
76824-368 76824-370 77611-636
Methyl 6-bromo-3-methoxypicolinate 98%
Methyl 6-bromo-3-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD18257515
CAS Number: 945954-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR