(2-Amino-5-(trifluoromethyl)phenyl)boronic acid 95% (by HPLC)

Supplier: Ambeed
Warning

A166949-1G A166949-250MG
76824-338EA 207.94 USD
76824-338 76824-340
(2-Amino-5-(trifluoromethyl)phenyl)boronic acid 95% (by HPLC)
(2-Amino-5-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₃NO₂
MW: 204.94 g/mol
MDL Number: MFCD07374868
CAS Number: 1793091-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR