3-Bromo-N,N-dimethylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A166247-1G A166247-5G
76824-312EA 75 USD
76824-312 76824-314
3-Bromo-N,N-dimethylpyridin-2-amine 95%
3-Bromo-N,N-dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD13188970
CAS Number: 1060801-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR