Methyl 6-chloro-1H-indole-3-carboxylate 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A166195-250MG
A166195-1G
76824-310EA
20
USD
76824-310
77683-363
Methyl 6-chloro-1H-indole-3-carboxylate 98%, Ambeed.Inc
Methyl 6-chloro-1H-indole-3-carboxylate
Precaución: For research use only.
Formula:
C₁₀H₈ClNO₂ MW: 209.63 g/mol |
MDL Number:
MFCD09836004 CAS Number: 921194-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...