Methyl 1-benzyl-4-hydroxypiperidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A165876-100MG A165876-1G A165876-250MG
76824-300EA 66.84 USD
76824-300 76824-302 76824-304
Methyl 1-benzyl-4-hydroxypiperidine-4-carboxylate 97%
Methyl 1-benzyl-4-hydroxypiperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.31 g/mol
MDL Number: MFCD17453986
CAS Number: 60437-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR