Methyl-2,5-difluorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,5-difluorobenzoate

A165861-100G A165861-10G A165861-1G A165861-25G A165861-5G
76824-290EA 168.69 USD
76824-290 76824-292 76824-294 76824-296 76824-298
Methyl-2,5-difluorobenzoate 97%
Methyl-2,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD03093800
CAS Number: 362601-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR