N-Benzyl-4-bromo-3-methylbenzamide 97%

Supplier: Ambeed
Warning

A165542-10G A165542-1G A165542-25G A165542-5G
76824-274EA 5.31 USD
76824-274 76824-276 76824-278 76824-280 76824-282 76824-284
N-Benzyl-4-bromo-3-methylbenzamide 97%
N-Benzyl-4-bromo-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrNO
MDL Number: MFCD09972121
CAS Number: 1020252-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR