(S)-2-Amino-2-cyclohexylethanol 98%

Supplier: Ambeed
Warning

A165044-1G A165044-250MG A165044-5G A165044-25G
76824-262EA 46 USD
76824-262 76824-264 76824-266 77885-309
(S)-2-Amino-2-cyclohexylethanol 98%
(S)-2-Amino-2-cyclohexylethanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MDL Number: MFCD04112589
CAS Number: 845714-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR