4-Bromo-6-iodo-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A164874-100MG
A164874-1G
A164874-250MG
A164874-5G
76824-244EA
103.97
USD
76824-244
76824-246
76824-248
76824-250
4-Bromo-6-iodo-1H-indazole 97%
4-Bromo-6-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄BrIN₂ MW: 322.93 g/mol |
MDL Number:
MFCD07781349 CAS Number: 885518-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...